Skip to Content
MilliporeSigma

STS0031

Sodium glycochenodeoxycholate

BioUltra, Used as detergent and surfactant

Synonym(s):

Bile acid salt, Trace element free Glycochenodeoxycholate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H42NNaO5
CAS Number:
Molecular Weight:
471.61
NACRES:
NA.21
UNSPSC Code:
12161900
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

General description

Glycochenodeoxycholate (GCDC) is a toxic hydrophobic bile salt, often abbreviated as NaGCDCA. It is a detergent and surfactant used in biochemical and molecular biology for protein solubilization and membrane protein purification. It has steroid structure containing both hydrophilic and hydrophobic regions, thus making them effective in solubilizing lipids and helping in digestion of facts in the gastro intestinal tract.

Application

Sodium glycochenodeoxycholate has been used as an additive in de Man, Rogosa and Sharpe (MRS) agar for bile acid deconjugation to determine the capacity of the strains to deconjugate specific bile acids. Sodium glycochenodeoxycholate has been used in a study to assess the repair of DNA damage induced by bile salts. It has also been used in a study to investigate enterotoxin production by Clostridium perfringens type A in the presence of human bile salts

Features and Benefits

Solubilize membrane proteins by disrupting lipid-lipid and lipid-protein interactions. Helps in isolating and studying membrane proteins.


Still not finding the right product?

Explore all of our products under Sodium glycochenodeoxycholate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library