Skip to Content
MilliporeSigma

STS0300

CALX8

Synonym(s):

5,11,17-Tris[(carboxy)methyl]-25-monooctyloxy-26,27,28-trihydroxycalix[4]arene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C42H43Na3O10
Molecular Weight:
776.75
NACRES:
NB.34
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Non-ionic

assay

95% (HPLC)

form

powder

manufacturer/tradename

CALIXAR®

CMC

2.2 mM

solubility

DMSO: soluble, methanol: soluble, water: soluble

storage temp.

−20°C

General description

CALX8 is a soft proprietary detergent. CALX8 is an ionic calixarene based detergent with three polar carboxylic groups and an octyl chain.

Application

CALX8 is used for

  • Extraction of membrane proteins
  • Solubilization of membrane proteins
  • Stabilization of membrane proteins

Features and Benefits

  • Mild detergent
  • Not sensitive to pH variations
  • Not sensitive to ionic strength
  • Able to be combined with biological materials
  • Can be used in a buffer or aqueous solution

Legal Information

CALIXAR is a registered trademark of CALIXAR SAS FRANCE


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library