Skip to Content
MilliporeSigma

STS0303

DDG

Synonym(s):

N-(2-Methyl-1,3-bis(O-ß-D-Glucose)propan-2-yl)-3-(decylthio)propanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H55NO13S
Molecular Weight:
657.81
NACRES:
NB.34
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Non-ionic

assay

95% (HPLC)

form

powder

manufacturer/tradename

CALIXAR®

CMC

0.54 mM

solubility

water: 27 mM, DMSO: soluble, methanol: soluble

storage temp.

−20°C

General description

DDG is a soft proprietary detergent. DDG is a non-ionic amphiphilic detergent and surfactant with a di-glucose polar group and decyl chain.

Application

DDG is used for
  • Extraction of membrane proteins
  • Solubilize membrane proteins
  • Stabilization of membrane proteins
  • Creating lipid detergents
  • Creating mixed micelles
  • Creating protein detergent micelles

Features and Benefits

  • Moderate detergent
  • Not sensitive to pH variations
  • Not sensitive to ionic strength
  • Able to be combined with biological materials
  • Can be used in a buffer or aqueous solution

Legal Information

CALIXAR is a registered trademark of CALIXAR SAS FRANCE


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library