Skip to Content
MilliporeSigma

STS0305

DLAC

Synonym(s):

N-Decylamine lactobionamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H43NO11
Molecular Weight:
497.58
NACRES:
NB.34
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Non-ionic

assay

97% (HPLC)

form

powder

manufacturer/tradename

CALIXAR®

CMC

1.3 mM

solubility

water: >100 mM, DMSO: soluble, methanol: soluble

storage temp.

−20°C

General description

DLAC is a soft proprietary detergent. DLAC is a non-ionic detergent with a lactobionamide head group and a decyl chain.

Application

DLAC is used for

  • Extraction of membrane proteins
  • Solubilization of membrane proteins
  • Stabilization of membrane proteins

Features and Benefits

  • Mild detergent
  • Not sensitive to pH variations
  • Not sensitive to ionic strength
  • Able to be combined with biological materials
  • Can be used in a buffer or aqueous solution

Legal Information

CALIXAR is a registered trademark of CALIXAR SAS FRANCE


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library