Skip to Content
MilliporeSigma

STS0308

DDTAC

Synonym(s):

Dodecylmercapto-S-(poly(tris(hydroxymethyl)acrylamidomethane) DPn=6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C54H104N6O24S
Molecular Weight:
1253.49
NACRES:
NB.34
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Non-ionic

form

powder

manufacturer/tradename

CALIXAR®

CMC

~0.15 mM

solubility

water: 15 mM, DMSO: soluble, methanol: soluble

storage temp.

−20°C

General description

DDTAC is a soft proprietary, non-ionic detergent.

Application

DDTAC is used for

  • Extraction of membrane proteins
  • Solubilization of fragile membrane proteins
  • Stabilization of fragile membrane proteins

Features and Benefits

  • Mild detergent
  • Not sensitive to pH variations
  • Not sensitive to ionic strength
  • Can be customized to solubilize and stabilize specific membrane proteins

Legal Information

CALIXAR is a registered trademark of CALIXAR SAS FRANCE


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library