Skip to Content
MilliporeSigma

T8938

Active Native Bovine Thyroid Stimulating Hormone

Lyophilized powder, ≥ 25IU/mg protein

Synonym(s):

TSH, Thyroid stimulating hormone, Thyrotropin from bovine pituitary glands

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
Biological source:
bovine pituitary glands
Form:
lyophilized powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pituitary glands

Quality Segment

form

lyophilized powder

mol wt

(α subunit ≈ 13–15 kDa
β subunit ≈ 15–17 kDa
Total ≈ 28–32 kDa)

purified by

chromatography

suitability

suitable for molecular biology

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

target post-translational modification

disulfide bond

Gene Information

General description

TSH is a heterodimeric glycoprotein hormone secreted by the anterior pituitary gland. It consists of a common α-subunit shared with other glycoprotein hormones (FSH, LH, and hCG) and a hormone-specific β-subunit that confers receptor specificity. Native bovine TSH is structurally conserved across mammalian species and exhibits biological activity through interaction with the thyroid-stimulating hormone receptor (TSHR). As a glycosylated protein, its biological activity and stability are influenced by its carbohydrate moieties.

Application

Thyroid stimulating hormone has been used in:
  • Studies of thyroid function and regulation
  • Cell-based assays involving TSH receptor activation
  • Endocrine signaling and hormone response studies
  • Assays evaluating thyroid hormone biosynthesis

Biochem/physiol Actions

TSH binds to the TSH receptor (TSHR) on thyroid follicular cells, activating cAMP-dependent signaling pathways that regulate thyroid hormone synthesis and secretion (T3 and T4). It plays a central role in maintaining metabolic homeostasis and regulating growth and development.

Physical form

Lyophilized from 10mM KPB (pH 8.0), 0.150M NaCl

Preparation Note

It is recommended that the powder be reconstituted in physiological saline (PH7.0)


Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library