Skip to Content

Z-2013

ZytoLight® SPEC MDM2/CEN 12 Dual Color Probe

green to orange, pkg of 0.2 mL, pkg of 0.05 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC MDM2/CEN 12 Dual Color Probe (PL18) is a direct‑labeled dual‑color fluorescence in situ hybridization (FISH) probe for detecting the human MDM2 gene region together with alpha‑satellite sequences of chromosome 12 in formalin‑fixed, paraffin‑embedded specimens.It combines differently labeled probe fractions targeting the MDM2 locus and chromosome 12 centromeric sequences to support visualization of copy number changes at these regions by FISH on tissue sections.

Principle:

This probe is intended for use in fluorescence in situ hybridization (FISH) procedures on formalin-fixed, paraffin-embedded tissue specimens. It is designed to support the analysis of genetic features and should be used alongside other relevant information and methods for comprehensive evaluation.

Components:Probe Composition:
  • ZyGreen: Labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting sequences in 12q15 (harboring the MDM2 gene).
  • ZyOrange: Labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences in 12p11.1-q11 (specific for the alpha satellite centromeric region D12Z3 of chromosome 12).
  • Hybridization Buffer: Formamide-based hybridization buffer.Related Products: Intended for use with the ZytoLight FISH-Tissue Implementation Kit (Prod. No. Z-2028-5/-20).

Application

This probe is intended for use in fluorescence in situ hybridization (FISH) procedures on formalin-fixed, paraffin-embedded tissue specimens. It is designed to support the analysis of genetic features and should be used alongside other relevant information and methods for comprehensive evaluation.
For the qualitative detection of human MDM2 gene amplifications and alpha satellites of chromosome 12 by fluorescence in situ hybridization (FISH)

Analysis Note

The performance of the probe has been validated for detecting amplifications of the MDM2 gene locus. In normal interphase cells, two green signals (representing the MDM2 gene region) and two orange signals (representing the centromeric region of chromosome 12) are expected. In cases of amplification, multiple copies of the green signal or clusters will be observed.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library