Skip to Content

Z-2078

ZytoLight® SPEC PTEN/CEN 10 Dual Color Probe

green to orange, pkg of 0.05 mL, pkg of 0.2 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC PTEN/CEN 10 Dual Color Probe (PL37) is a direct-labeled fluorescence in situ hybridization (FISH) probe set containing green and orange labeled polynucleotides targeting sequences in the PTEN gene region and chromosome 10 alpha satellite centromeric region in formalin-fixed, paraffin-embedded (FFPE) tissue specimens. This probe utilizes FISH technology for the visualization of specific nucleic acid sequences.

Principle:

Fluorescence in situ hybridization (FISH) detects and visualizes specific nucleic acid sequences in cell preparations. Fluorescently-labeled DNA fragments (FISH probes) co-denature and hybridize with complementary target DNA strands, followed by stringent washing to remove unbound probes. The hybridized probes are visualized using a fluorescence microscope with specific excitation and emission filters.FISH assesses the copy number of PTEN and CEN10 using direct probes that hybridize after denaturation, with unbound probes washed away and a DAPI counterstain applied. The signals—green for PTEN and orange for CEN10—indicate deletions. This antibody-free method is optimized for fluorochromes, aiding in the identification of tumor suppressor gene aberrations.

Components:

ZytoLight® SPEC PTEN/CEN 10 Dual Color Probe comprises:

  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting the PTEN gene region on chromosome 10 (10q23.2-q23.31).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting the alpha satellite centromeric region D10Z1 of chromosome 10.k
  • Formamide-based hybridization buffer.

Application

For the qualitative detection of human PTEN gene deletions and alpha satellites of chromosome 10 by fluorescence in situ hybridization (FISH)
The probe is designed for qualitative detection of sequences in the PTEN gene region and chromosome 10 centromere in formalin-fixed, paraffin-embedded (FFPE) tissue specimens by FISH. It is used in combination with the ZytoLight® FISH-Tissue Implementation Kit for specimen pretreatment, hybridization, and post-hybridization processing.

Analysis Note

In a normal interphase nucleus, the expected result is the presence of two orange and two green signals. If there is a deletion of the PTEN gene locus, a reduced number of green signals will be observed. Variations in signal patterns may indicate different genetic aberrations and should be investigated further. Proper specimen handling and processing are crucial to avoid artifacts or false results.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library