packaging
pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests
storage condition
protect from light
color
green to orange
application(s)
research use
storage temp.
2-8°C
General description
The ZytoLight SPEC RET Dual Color Break Apart Probe (PL105) is a direct-labeled FISH probe designed for the qualitative detection of translocations involving the human RET gene at 10q11.21 in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended to be used with the ZytoLight FISH-Tissue Implementation Kit.
Components:
ZytoLight® SPEC RET Dual Color Break Apart Probe comprises:
Components:
ZytoLight® SPEC RET Dual Color Break Apart Probe comprises:
- ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting sequences distal to the RET breakpoint region on chromosome 10 (10q11.21).
- ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences proximal to the RET breakpoint region.
- Formamide-based hybridization buffer.
Application
For the qualitative detection of translocations involving the human RET gene at 10q11.21 by fluorescence in situ hybridization (FISH)
ZytoLight® SPEC RET Dual Color Break Apart Probe is designed for the qualitative assessment of rearrangements involving the RET gene locus in formalin‑fixed, paraffin‑embedded human tissue sections using fluorescence in situ hybridization (FISH). It is intended for use by trained personnel in appropriately equipped laboratories as part of broader histopathological and molecular investigations, with results to be interpreted in conjunction with additional laboratory findings and relevant clinical information.
Analysis Note
In a normal interphase nucleus, two green signals (for RET) and two orange signals (for the RET breakpoint region) are expected. In cases of translocation, separate signals will be observed. Signal patterns should be interpreted carefully to identify potential genetic rearrangements.
Legal Information
ZytoLight is a registered trademark of ZytoVision GmbH in Germany
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 1B - STOT RE 2 Oral
Target Organs
Blood
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
