Skip to Content

Z-2161

ZytoLight® SPEC ALK/2q11 Dual Color Probe

green to orange, pkg of 0.2 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.2 mL, pkg of 20 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight SPEC ALK/2q11 Dual Color Probe (PL117) is a direct-labeled FISH probe designed for the qualitative detection of amplifications involving the human ALK gene and the detection of chromosome 2q11 specific sequences in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended to be used with the ZytoLight FISH-Tissue Implementation Kit.

Components:

ZytoLight® SPEC ALK/2q11 Dual Color Probe comprises:

  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting the ALK gene region on chromosome 2 (2p23.1).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences specific to chromosome 2q11.
  • Formamide-based hybridization buffer.
Principle:

Fluorescence in situ hybridization (FISH) uses direct-labeled probes (ALK, 2q11) that co-denature with target DNA in pretreated FFPE sections (10 µl/slide, sealed), hybridize overnight (75°C/10 min denaturation, 37°C hybridization), then undergo stringent washing and DAPI counterstaining for visualization via fluorescence microscopy with color-specific filters per ZytoLight FISH-Tissue Implementation Kit.

Application

The ZytoLight® SPEC ALK/2q11 Dual Color Probe is used to detect RET gene rearrangements and ALK copy number gains/amplifications in formalin-fixed, paraffin-embedded tissue specimens, to support molecular characterization within the analytical workflow. In addition to RET rearrangement detection, ALK copy number alterations may be observed in a variety of tissue types, providing information about gene copy status within the context of the assay.ance.

Analysis Note

In a normal interphase nucleus, two green signals (for ALK) and two orange signals (for the 2q11 locus) are expected. In cases of amplification, an increased number of green signals or clusters will be observed. Any unexpected signal patterns should be further investigated for potential genetic rearrangements.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library