packaging
pkg of 0.2 mL, pkg of 20 tests
storage condition
protect from light
color
green to orange
application(s)
research use
storage temp.
2-8°C
General description
The ZytoLight SPEC ROS1/CEN 6 Dual Color Probe (PL118) is a direct-labeled FISH probe intended for the qualitative detection of amplifications involving the human ROS1 gene and for detecting chromosome 6 alpha satellites in formalin-fixed, paraffin-embedded (FFPE) specimens. It is designed for use in conjunction with the ZytoLight FISH-Tissue Implementation Kit.
Components:
ZytoLight® SPEC ROS1/CEN 6 Dual Color Probe comprises:
Fluorescence in situ hybridization (FISH) uses direct-labeled probes (ROS1, CEN 6) that co-denature with target DNA in pretreated FFPE sections (10 µl/slide, sealed), hybridize overnight (75°C/10 min denaturation, 37°C hybridization), then undergo stringent washing and DAPI counterstaining for visualization via fluorescence microscopy with color-specific filters per ZytoLight FISH-Tissue Implementation Kit.
Components:
ZytoLight® SPEC ROS1/CEN 6 Dual Color Probe comprises:
- ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting sequences in the ROS1 gene region on chromosome 6 (6q22.1).
- ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting the alpha satellite centromeric region D6Z1 of chromosome 6.
- Formamide-based hybridization buffer.
Fluorescence in situ hybridization (FISH) uses direct-labeled probes (ROS1, CEN 6) that co-denature with target DNA in pretreated FFPE sections (10 µl/slide, sealed), hybridize overnight (75°C/10 min denaturation, 37°C hybridization), then undergo stringent washing and DAPI counterstaining for visualization via fluorescence microscopy with color-specific filters per ZytoLight FISH-Tissue Implementation Kit.
Application
ZytoLight® SPEC ROS1/CEN 6 Dual Color Probe is used to detect ROS1 gene amplifications and alpha satellite regions on chromosome 6, aiding in differential analyses where ROS1 alterations are relevant. The probe is designed to be used in conjunction with the ZytoLight FISH-Tissue Implementation Kit (Z-2028-5/-20).
Analysis Note
In a normal interphase nucleus, two green signals (for ROS1) and two orange signals (for chromosome 6) are expected. In cases of amplification or translocation, an increased number of green or orange signals may be observed, which should be further investigated for potential genetic rearrangements.
Legal Information
ZytoLight is a registered trademark of ZytoVision GmbH in Germany
Target Organs
Blood
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 1B - STOT RE 2 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
