Skip to Content

Z-2169

ZytoLight® SPEC FGFR2 Dual Color Break Apart Probe

green to orange, pkg of 0.2 mL, pkg of 0.05 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

Application

For the qualitative detection of translocations involving the FGFR2 gene at 10q26.13 by fluorescence in situ hybridization (FISH)

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library