Skip to Content

Z-2173

ZytoLight®SPEC MCL1/1p12 Dual Color Probe

green to orange, pkg of 0.2 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.2 mL, pkg of 20 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight SPEC MCL1/1p12 Dual Color Probe (PL129) is a direct-labeled FISH probe intended for the qualitative detection of amplifications involving the human MCL1 gene and for detecting specific sequences on chromosome 1p12 in formalin-fixed, paraffin-embedded (FFPE) specimens. It is designed to be used in conjunction with the ZytoLight FISH-Tissue Implementation Kit.

ZytoLight®SPEC MCL1/1p12 Dual Color Probe comprises:
  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting the MCL1 gene region on chromosome 1 (1q21.3).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences specific to chromosome 1p12.
  • Formamide-based hybridization buffer.
Principle:
FISH detects specific sequences via direct-labeled probes co-denatured (75°C/10 min) with target DNA in pretreated FFPE sections (10 µl/slide), hybridized overnight (37°C), stringency washed, DAPI counterstained, and visualized by fluorescence microscopy per ZytoLight FISH-Tissue Implementation Kit (Z-2028-5/-20).

Application

For the qualitative detection of human MCL1 gene amplifications and chromosome 1p12 specific sequences by fluorescence in situ hybridization (FISH)

Analysis Note

In a normal interphase nucleus, two orange and two green signals are expected. In cases of MCL1 amplification, multiple green signals or clusters will be observed. Unexpected signal patterns should be investigated further for potential genetic rearrangements.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library