Skip to Content

Z-2176

ZytoLight® SPEC ETV6 Dual Color Break Apart Probe

green to orange, pkg of 0.2 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight SPEC ETV6 Dual Color Break Apart Probe (PL135) is a direct-labeled FISH probe designed for the qualitative detection of translocations involving the human ETV6 gene at 12p13.2 in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended for use with the ZytoLight FISH-Cytology Implementation Kit.

Components:

ZytoLight® SPEC ETV6 Dual Color Break Apart Probe comprises:

  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting sequences proximal to the ETV6 breakpoint region on chromosome 12 (12p13.2).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting sequences distal to the ETV6 breakpoint region.
  • Formamide-based hybridization buffer.
Principle:
FISH employs direct-labeled probes flanking ETV6 breakpoint, co-denatured (72°C/5 min) with target DNA on pretreated cytology slides (10 µl/slide), hybridized overnight (37°C), stringency washed, DAPI counterstained, and visualized by fluorescence microscopy using ZytoLight FISH-Cytology Implementation Kit (Z-2099-20).

Application

ZytoLight® SPEC ETV6 Dual Color Break Apart Probe aids in the differential analysis of conditions associated with ETV6 rearrangements, including ESS and other related contexts. It is used for the qualitative detection of translocations involving the ETV6 gene at 12p13.2 by fluorescence in situ hybridization (FISH)

Analysis Note

In a normal interphase nucleus, two green and two orange signals are expected. In cases of ETV6 rearrangements, an aberrant signal pattern may present as one orange/green fusion signal, one orange signal, and one separate green signal, indicating the presence of a translocation. Any unexpected signal patterns should be further investigated.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library