MilliporeSigma
Z355305
Millipore® Sterivex™ filter unit
HV with luer-lock outlet, pore size 0.45 μm, Durapore PVDF membrane
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.24
UNSPSC Code:
41104900
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
Durapore PVDF membrane, housing (Eastar)
sterility
sterile
packaging
pack of 15
manufacturer/tradename
Millipore SVHVL10RC
pore size
0.45 μm
General description
Suitable for sterilizing and clarifying tissue culture media and other aqueous solutions in the 100 mL to 1 liter volume range. Pressure-driven, self-venting, direct-dispensing design allows filtration into any type of container without foaming. Can be used with a peristaltic pump, pressure vessel, or syringe.
Connections are female luer-lock inlet and 3.3 mm O.D. male tubing outlet or male luer-lock outlet.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Sterivex is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)