Skip to Content
MilliporeSigma

Z741865

Cameo™ Syringe Filter

cellulose acetate membrane, pore size 0.22 μm, diam. 30 mm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41104900
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cellulose acetate membrane, polypropylene housing

sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

GVS Life Sciences 1213641

diam.

30 mm

pore size

0.22 μm

General description

Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.

Applications:
Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis

Legal Information

Cameo is a trademark of GVS S.p.A.


Still not finding the right product?

Explore all of our products under Cameo™ Syringe Filter




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library