Skip to Content
MilliporeSigma

Z744252

Wilmad® 7 mm Bruker® MAS Rotor Set

includes 2 rotors, 2 Kel-F′ Caps, 1 Torlon® cap

Synonym(s):

Solid State NMR, MAS NMR, Solid State NMR, MAS NMR,, NMR rotor set, Wilmad LabGlass WP-501-7180-SET-2, wilmad NMR rotor set

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
23152100
Manufacturer/tradename:
Wilmad WP-501-7180-SET-2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 1 ea (2 rotors, 2 Kel-F Caps, 1 Torlon cap)

manufacturer/tradename

Wilmad WP-501-7180-SET-2

General description

Manufactured from the highest purity Zirconia, MAS Rotors provide the NMR spectroscopist with the ultimate alternative for analysis of solid samples; the need to solvate is eliminated. They are available for most present solid state NMR spectrometers. MAS Rotors are carefully examined for material irregularities by optical methods before and after the precision machining process. Without over-spinning, each rotor is spin tested to only the highest specified spinning speed. Overspin testing, a common practice with some rotor brands, can actually damage the rotor prior to use. High precision (which becomes a necessity for proper spinning performance) is maintained in the manufacture of the end caps. Most caps are fitted with O-rings for better sealing; some have axial holes for venting. Advantages of Zirconia Rotors Most homogeneous ceramic material structure. Zirconia Rotor Body has higher strength (1,000 MPa, greater than Si3N4). Highest spinning speed, high precision superior finish (0.05 µ mirror gloss). Some caps have a uniquely designed detachable O-ring for use when a gas-tight environment is required for the sample.

Application

NMR, MAS NMR

Features and Benefits

  • Homogeneous ceramic material structure.
  • Zirconia Rotor Body strength 1,000 MPa
  • Finish 0.05 µ mirror gloss

Legal Information

Bruker is a registered trademark of Bruker-Physik AG
Torlon is a registered trademark of Amoco Chemicals Corporation
Wilmad is a registered trademark of SP Industries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library