Skip to Content
MilliporeSigma

Z744286

Wilmad® Vespel™ Variable Temperature CFQ Transfer Dewar for Bruker®

OAL, L 725 mm, pack of 1 ea

Synonym(s):

EPR cryo dewar, EPR dewar, Wilmad LabGlass WG-842-MX, wilmad dewar, wilmad liquid nitrogen dewar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.15
UNSPSC Code:
41103800
Packaging:
pack of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-842-MX

L

725 mm

General description

Longer version transfer line designed for Bruker ER4121 VT systems in an EPR console with magnet >18 cm diameter.Dim. A (mm):725Dim. B (mm):75Dim. C (mm):110

Application

EPR

Legal Information

Bruker is a registered trademark of Bruker-Physik AG
Vespel is a trademark of E. I. du Pont de Nemours and Company
Wilmad is a registered trademark of SP Industries




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library