Skip to Content
MilliporeSigma

Z744310

Multi-Coloured Well Oritenter

wells, 384, hotizontal, pack of 2 ea

Synonym(s):

Diversified Biotech WELL-384H, cell culture plate alignment tool, multi-well aligner, multi-well positioner, well alignment tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122102
Manufacturer/tradename:
Diversified Biotech WELL-384H
Packaging:
pack of 2 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

non-sterile

feature

wells 384, hotizontal

packaging

pack of 2 ea

manufacturer/tradename

Diversified Biotech WELL-384H

General description

For easy identification of samples in multiwell plates, available in 96 or 384 well formats, in either horizontal or vertical orentintation and colour coded to aide ease of use. Can be used a pipette guide for convenient viewing.

Application

Cell Culture

Features and Benefits

  • Fits most standard plates
  • Easy to use



Still not finding the right product?

Explore all of our products under Multi-Coloured Well Oritenter




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library