Skip to Content
MilliporeSigma

Z744314

Well Orienter Stickers

wells, 96, pack of 50 ea

Synonym(s):

Diversified Biotech WEST-1000, cell culture plate alignment tool, multi-well aligner, multi-well positioner, well alignment tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122102
eCl@ss:
32150313
Manufacturer/tradename:
Diversified Biotech WEST-1000
Material:
acrylic (cardboard backing), clear (black text)
Packaging:
pack of 50 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

acrylic (cardboard backing), clear (black text)

sterility

non-sterile

feature

wells 96

packaging

pack of 50 ea

manufacturer/tradename

Diversified Biotech WEST-1000

General description

Permanently label mutliple microplates with clear Well Orienter Stickers. Each well is clearly marked with its corresponding row number and column letter for easy cross-referencing. The paper backing is scored in the middle allowing for easy aligning on the underside of the plate.

Application

Cell Culture




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library