MilliporeSigma
Z744324
Vacuubrand Inlet separator AK for VACUU·PURE
round bottom flask 500ml, KF DN 25, hose nozzle DN15 mm
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20751802
General description
Inlet separator AK with round bottom flask 500 ml, with KF DN 25, for VACUU·PURE Inlet separator for VACUU·PURE Screw pumps. Inlet with small flange KF DN 25 and hose nozzle DN 15 mm. Separators (AK) protect the vacuum pump at the inlet from particles and liquid droplets. Separators catchpot made of glas with good chemical resistance.
Application
protect screw pumps from particles and liquid droplets
Features and Benefits
For VACUU·PURE Screw Pumps
  • Direct mounting at the inlet, compact and leak-tight
  • High conductance
  • Direct visibility of condensate through the coated round bottom flask
  • Easy draining of condensate
Choose from one of the most recent versions:
Certificates of Analysis (COA)