MilliporeSigma
Z744329
Vacuubrand KF DN 25/SW10, PP
small flange with hose nozzle
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20662807
General description
Small flange with a hose nozzle, made of polypropylene (PP), designed for a DN 25 (Nominal Diameter 25 mm) KF (Klein Flange) connection and a hose barb with a 10mm outer diameter (SW10)
Choose from one of the most recent versions:
Certificates of Analysis (COA)