MilliporeSigma
Z744340
Vacuubrand set of connection parts for VHCpro
with VHCpro hose connection and inlet tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
eCl@ss:
32039090
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20699943
General description
Extension kit second VHC connection. Hose nozzle and inlet tube for minimized foam and aerosol formation (VHC / VHCpro not included). For BVC professional with 4l bottle PP please order article Z744347 instead
Application
fluid aspiration accessories
Choose from one of the most recent versions:
Certificates of Analysis (COA)