MilliporeSigma
Z744341
Vacuubrand BVC shuffle
mobile underframe for BVC control and BVC professional
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
eCl@ss:
32150313
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20696880
General description
The BVC shuttle is a mobile underframe for all BVC control and BVC professionalmodels. It is used to easily pull the BVC out from under the safety cabinet forcleaning and changing bottles.
The BVC shuttle is a mobile underframe for all models BVC control and BVC professional. It serves to pull out the BVC easily from under the safety cabinet for cleaning and removal of bottle for change
Application
fluid aspiration accessories
Features and Benefits
Enables easy access to BVC from under the safety cabinet forcleaning and changing bottles
Choose from one of the most recent versions:
Certificates of Analysis (COA)