MilliporeSigma
Z744346
Vacuubrand 4L bottle for BVC system
polypropylene, with hose nozzle for connection to the VHC system
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.17
Capacity:
-4 L
Material:
polypropylene
Sterility:
non-sterile
Feature:
with cap
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
non-sterile
feature
with cap
manufacturer/tradename
Vacuubrand 20635810
capacity
-4 L
General description
4l collection bottle with high chemical resistance (PP). Special bottle capfor minimized aerosol and foam formation. Hose nozzle for connection ofa VacuuHandControl VHC / VHCpro. Additional port to connect a secondVHCpro (optional). Protective filter 0.2 μm with hydrophobic membrane toprotect pump, lines and environment with hose for connecting to a BVC.Completely autoclavable.
4L heavy-walled, vacuum-proof collection bottle with high chemical resistance (PP). Special bottle head for minimized foam and aerosol formation, with hose nozzle for VacuuHandControl VHC / VHCpro and additional port to connect a second VHC / VHCpro (optional). Hydrophobic filter element 0.2 µm to protect pump and environment. Hose for connection to BVC. Fully autoclavable.
Application
fluid aspiration accessories
Features and Benefits
  • High chemical resistance
  • Special bottle capfor minimized aerosol and foam formation
  • Hose nozzle for connection ofa VacuuHandControl VHC / VHCpro with additional port to connect a secondVHCpro (optional)
  • Protective filter 0.2 μm with hydrophobic membrane toprotect pump, lines and environment with hose for connecting to a BVC
  • Completely autoclavable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)