MilliporeSigma
Z744347
Vacuubrand quick coupling VHC/VHCpro to collection bottle for BVC system
PVDF, chemical resistant, autoclavable
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20635807
General description
Set of quick-coupling for connection VHC-bottle including adapter and inlet tube. Quick coupling made of PVDF, with adapter to connect a VHC / VHCpro to a collection bottle. Very good chemical resistance, complete with inlet tube for minimized foam and aerosol formation. Autoclavable. This set is also suitable to connect a second VHC/VHCpro. When disconnected, the collection bottle closes vacuum tight.
Complete with quick connect fitting and male quick coupling, adapter and inlet tube.
Application
fluid aspiration accessories
Features and Benefits
  • Quick coupling made of PVDF for exceptional chemical resistance
  • Complete with inlet tube for minimized foam and aerosol formation
  • Autoclavable
  • Suitable to connect a second VHC/VHCpro
  • When disconnected, the collection bottle closes vacuum tight
Choose from one of the most recent versions:
Certificates of Analysis (COA)