MilliporeSigma
Z744348
Vacuubrand Table stand for VHCpro
stainless steel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
eCl@ss:
32150313
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20635680
General description
The stainless steel stand for the VHCpro enables to place the tipped suction handset conveniently and stable during work with cell cultures. It is autoclavable and comes with slip-resistant feet. The stand supports to place the VHCpro also with long, e. g. graduated pipettes, without touching the working surface. The stainless steel stand for the VHCpro enables to place the tipped suction handset conveniently and stable during work with cell cultures
Application
fluid aspiration accessories
Choose from one of the most recent versions:
Certificates of Analysis (COA)