MilliporeSigma
Z744349
Vacuubrand 8-channel adapter for pipette tips with ejector
for BVC system. Tips size 2-200µl or 5-300µl.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.01
eCl@ss:
32079201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20635679
General description
This adapter allows parallel operation of eight pipette tips (size 2 - 200 µl or 5 - 300 µl). It is designed to match the dimensions of standardized 96-well microplates and has got a tip ejector to remove the used pipette tips.
Application
fluid aspiration accessories
Features and Benefits
  • Suction power is evenly spread across all pipette tips
  • Completely autoclavable
  • Stand and disinfectant reservoir inclusive
  • Low effort for attaching and ejecting the tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)