MilliporeSigma
Z744350
Vacuubrand quick coupling to connect bottle to pump
for BVC system. PVDF, chemical resistant.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
eCl@ss:
32040102
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20635808
General description
Quick coupling made of PVDF, to connect a collection bottle to a BVC unit. Very good chemical resistance. Autoclavable. When disconnected, the collection bottle closes vacuum tight Quick coupling made of PVDF, to connect a collection bottle to a pump unit BVC or to a BVC basic, very good chemical resistance, autoclavable. When disconnected, the collection bottle closes vacuum tight
Features and Benefits
fluid aspiration accessories
Choose from one of the most recent versions:
Certificates of Analysis (COA)