MilliporeSigma
Z744351
Vacuubrand gauge VACUU·VIEW extended
100-230 V/50-60 Hz, plug CEE/CH/UK/US/AUS
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
40151700
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Vacuubrand 20683210
General description
For the monitoring of your vacuum processes in the rough and fine vacuum range, the new vacuum gauges VACUU·VIEW extended are perfect! The heavy duty combination of ceramic diaphragm sensor and ceramic jacketed Pirani sensor ensures reliable readings in the wide range from atmosphere down to 10-3 mbar.
Features and Benefits
  • Precision and chemical resistance in an exceptionally wide range from atmosphere down to 10-3 mbar, one gauge only for both rough and fine vacuum
  • Compact design with integrated sensors for useful setup in laboratory and process
  • The chemical resistant combination of ceramic diaphragm sensor and ceramic jacketed Pirani sensor ensures a long product life even with aggressive chemicals
  • Illuminated display, easy to read
  • Display with menu driven handling, easy to use e.g. for unit settings
Choose from one of the most recent versions:
Certificates of Analysis (COA)