Skip to Content
MilliporeSigma

Z744487

ClearASeal Pierce™ heat sealable film

LabSheet™, W × L 78 mm × 125 mm, sterile, pkg of 100 ea (pack of 100 sheets)

Synonym(s):

PCR storage, pierceable film, plate seal, qPCR storage, IST Scientific IST-103-078SS, film closure, laboratory sealing film, sealing film

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122410
NACRES:
NB.35
eCl@ss:
32120613
Manufacturer/tradename:
IST Scientific IST-103-078SS
W × l:
78 mm × 125 mm
Sterility:
sterile
Packaging:
pkg of 100 ea (pack of 100 sheets)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile

feature

piercable

packaging

pkg of 100 ea (pack of 100 sheets)

manufacturer/tradename

IST Scientific IST-103-078SS

technique(s)

PCR: suitable, qPCR: suitable

W × L

78 mm × 125 mm

suitability

suitable for PCR, suitable for fluorescence

General description

ClearASeal Pierce™ is a thin polyester, optically clear polyester backed film, forming a pierceable seal to polypropylene, polyethylene, polystyrene and cyclic olefin copolymer (COC) plates. The optical clarity of this seal enables its use for sealing plates required for imaging use, including fluorescent detection methods such as qPCR and colorimetric assays. It forms a complete seal to a plate enabling moderately low and high temperature uses, including PCR when using a pressurized heated lid thermal cycler and demonstrates a moderate solvent resistance and can be utilized for short term compound storage. This seal is available as sheets, for use with manual and semi-automated sealers and Its pierceability renders it useful for automation and for use on needle, capillary and tip based liquid handling systems It is effective on the ABI®3730 capillary sequencer, removing the need for the use of expensive septa mats.

Application

Suitable for PCR and qPCR, and short term storage

Features and Benefits

  • Permanent seal
  • Pierceable
  • Seal integrity range: -80 °C to 80 °C (or 110 °C when used with pressurized heated PCR lid)
  • Moderate solvent resistance
  • Optically clear

Legal Information

ClearASeal Pierce is a trademark of iST Scientific
LabSheet is a trademark of IST Scientific


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library