Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
eCl@ss:
32120613
Manufacturer/tradename:
IST Scientific IST-107-078LS
W × l:
78 mm × 125 mm
Sterility:
non-sterile
Packaging:
pkg of 100 ea
sterility
non-sterile
feature
piercable
packaging
pkg of 100 ea
manufacturer/tradename
IST Scientific IST-107-078LS
technique(s)
PCR: suitable, qPCR: suitable
W × L
78 mm × 125 mm
suitability
suitable for PCR
General description
PierceASeal Foil™ PS Heat Seal produces a strong seal to polystyrene plates and is compatible with polypropylene and polystyrene plates. This seal demonstrates moderate solvent resistance and can be used for low temperature compound storage, in DMSO and organic solvents, and short term room temperature storage. PierceASeal Foil PS can be pierced with a pipette tip manually, by a liquid handling robot, or it can be removed by peeling (from polystyrene only). It can be resealed by applying another Polystyrene Foil Heat Seal directly on top of a previously pierced seal. This seal is available as sheets, for use with manual and semi-automated sealers.
Application
- PCR
- Short term room temperature storage
- Low temperature storage
Features and Benefits
- Pierceable
- Seals polypropylene and polystyrene plates
Legal Information
LabSheet is a trademark of IST Scientific
PierceASeal Foil is a trademark of IST Scientific
Still not finding the right product?
Explore all of our products under PierceASeal Foil™ PS heat sealable film
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.