Skip to Content
MilliporeSigma

Z744583

Round-tip Thirsty-Swabs

pkg of 100 ea (swabs per pk)

Synonym(s):

swab kit, swab kits, swabs, Diversified Biotech SWAB-1000, absorbant wick

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41111700
NACRES:
NB.82
eCl@ss:
32049090
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 100 ea (swabs per pk)

manufacturer/tradename

Diversified Biotech SWAB-1000

General description

Round-tip Thirsty-Swabs, 100/pk

Application

Microscopy


Still not finding the right product?

Explore all of our products under Round-tip Thirsty-Swabs




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library