Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
Heathrow Scientific 120962
Feature:
to hold up to 200 x tubes between 10-30mm
Material:
PETG (thermoformed)
Packaging:
pack of 3
material
PETG (thermoformed)
feature
to hold up to 200 x tubes between 10-30mm
packaging
pack of 3
manufacturer/tradename
Heathrow Scientific 120962
L × W × D
13.1 in. × 14.3 in. × 5.6 in.
color
clear
General description
Designed to fit Duraporter® XL High-Capacity Sample Transport
Application
Can be used to securely hold tubes and samples.
Features and Benefits
- Holds tube with various diameters: 13, 18 & 30 mm, most 15 mL & 50 mL conical tubes, and 3.5 mL - 10 mL blood tubes
- Tube Rack Inserts can be used separately
- Lid removes completely for easy cleaning
- Insert deep enough to be filled with ice
Legal Information
Duraporter is a registered trademark of Heathrow Scientific
Still not finding the right product?
Explore all of our products under DuraPorter® XL Tube Rack
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.