MilliporeSigma
Z744740
Ground Glass Flask Stoppers
clear, joint: No. 16
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122502
Material:
glass
Color:
clear
Packaging:
pkg of 6 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
glass
joint
joint: No. 16
packaging
pkg of 6 ea
manufacturer/tradename
Heathrow Scientific 03083
color
clear
General description
The SIBATA Glass Stopper is lightweight yet very strong to reduce chipping and breakage. It meets ASTM E-675 specifications.
Features and Benefits
  • Flat top design allows the stopper to stand on its head, freeing the user′s hands
  • Fully interchangeable with other common flask stoppers and fit all standard labware of comparable standard taper size
  • Tinting: Clear
  • Standard Taper Stopper No.: 16
Still not finding the right product?
Explore all of our products under Ground Glass Flask Stoppers
Choose from one of the most recent versions:
Certificates of Analysis (COA)