MilliporeSigma
Z744792
PCR Rack 32 Place
blue, pack of 10
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122804
Manufacturer/tradename:
Heathrow Scientific 120889
Packaging:
pack of 10
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 10
manufacturer/tradename
Heathrow Scientific 120889
color
blue
General description
Designed for pre- and post-PCR sampling, streamline your PCR workflow with the 32-Place PCR Rack. This versatile rack accommodates 0.2 mL tubes both individually and in strips of 8, as well as 32-place assay plates (8x4)
Features and Benefits
Space Saving Design
  • Small and compact design is stackable, saving valuable space in the freezer
  • Right sized when processing smaller batches
  • Lid hinge design allows lid to to sit at 180 degrees, clear of sample processing, or can be removed entirely
  • Reusable Convenience
  • Transparent lid protects samples and allows for quick assessment of contents
  • Embossed numbers and letters on the wells provide easy identification andtracking of samples
  • Light-weighted construction made durable for reusability
Choose from one of the most recent versions:
Certificates of Analysis (COA)