MilliporeSigma
Z744800
Refractometer Standard
Water-based, 0.00% Brix 1.33297 nD (20°C), 7.4 mL/0.25 oz
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
51191600
eCl@ss:
32160703
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 45000-OS200
General description
Refractometer Standard, Oil-based, 60.0% Brix 1.4400 nD ± 0.0002 (25.0°C), 7.4 mL/0.25 oz Letter of NIST Traceability Certification.
Still not finding the right product?
Explore all of our products under Refractometer Standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)