MilliporeSigma
Z744802
Replacement Daylight Plate Kit
for Model BTX-20 refractometer
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 1 ea
manufacturer/tradename
Heathrow Scientific 43000-DLBTX20
General description
Replacement Daylight Plate Kit for handheld refractometer.
Application
Daylight plates are used to ensure accurate and consistent readings of the refractive index, especially when working with transparent liquids or samples that are challenging to read under standard lighting conditions.
Features and Benefits
  • Fits Model BTX-20 refractometer
  • Kit includes: Ring, Hing, Plate, and Screw
  • Assembled
Still not finding the right product?
Explore all of our products under Replacement Daylight Plate Kit
Choose from one of the most recent versions:
Certificates of Analysis (COA)