Skip to Content
MilliporeSigma

Z745201

TWD BioX Tissue Culture Flasks

300 cm2, rectangular canted neck with vented cap, 850ml, sterile, γ-irradiated

Synonym(s):

cell culture flask, graduated culture flask, plastic culture flask, sterile culture flask

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122104
eCl@ss:
32040201
Binding type:
Tissue Culture (TC)-treated surface
Material:
rectangular polystyrene flask (canted neck)
Packaging:
case of 18
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

rectangular polystyrene flask (canted neck)

packaging

case of 18

manufacturer/tradename

TWD Trade Winds Inc. BX078300

binding type

Tissue Culture (TC)-treated surface

General description

BioX Sterile Tissue Culture Flask with vented cap are designed for contamination-free cell growth in laboratory applications.

Application

Cell Culture

Features and Benefits

  • Flat surface free from striation to maximize growth area
  • Vented polythylene caps contain 0.22um hydrophobic filter allows gas exchange and minizes risk of cross-contamination
  • Engraved graduations on flank sides
  • Sterilized by gamma radiation, non-pyrogenic
  • TC-treated



Still not finding the right product?

Explore all of our products under TWD BioX Tissue Culture Flasks




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library