Skip to Content
MilliporeSigma

Z745207

TWD BioX Tissue Culture Plates

96 well, round bottom, polystyrene, sterile, individually wrapped

Synonym(s):

96-well plate, cell culture plate, individually wrapped plate, sterile plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122102
eCl@ss:
32150313
Material:
polystyrene
Size:
96 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

packaging

case of 100

manufacturer/tradename

TWD Trade Winds Inc. BX076096U

size

96 wells

binding type

Tissue Culture (TC)-treated surface

General description

BioX Sterile Tissue Culture TC-treated plates with coded wells for easy identification. Made of Polystyrene.

Application

Cell Culture

Features and Benefits

  • Smooth surface free from striation to maximize growth area
  • Raised rims on the wells with uniform rings on the lid to minimize evaporation
  • Single-position lid reduces risk of cross-contamination and handling errors
  • Wells are labelled with alphanumeric code for easy identification
  • Individually wrapped, sterile


Still not finding the right product?

Explore all of our products under TWD BioX Tissue Culture Plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library