Skip to Content
MilliporeSigma

Z745211

TWD BioX Serological Pipettes

10 mL, polystyrene, sterile, DNAse/Rnase-free, individually wrapped

Synonym(s):

10mL serological pipettes, pipettes for cell culture, serological pipets, sterile pipettes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121511
eCl@ss:
32070103
Packaging:
case of 200 (Individually wrapped)
Feature:
DNAse-free; RNAse-Free
Manufacturer/tradename:
TWD Trade Winds Inc. BX075010
Material:
polystyrene
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

feature

DNAse-free; RNAse-Free

packaging

case of 200 (Individually wrapped)

manufacturer/tradename

TWD Trade Winds Inc. BX075010

volume

10 mL

General description

BioX serological pipettes are available in a range of sizes, from 1mL to 50mL. These pipettes feature a clear, graduated markings for precise volume measurements. Made of polystyrene.

Application

Cell Culture

Features and Benefits

  • Volumetric accuracy: ±2% at full volume
  • Individually wrapped with paper/plastic
  • Sterile, non-pyrogenic, DNase/RNase-free



Still not finding the right product?

Explore all of our products under TWD BioX Serological Pipettes




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library