Skip to Content
MilliporeSigma

ZRB3081

Anti-CD200 Antibody, clone 1F7 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells

Sustainability features

enhanced validation

Enhanced Validation

Synonym(s):

MOX1, MOX2, My033, OX-2 membrane glycoprotein, OX2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1F7, recombinant monoclonal
Application:
IHC, WB, affinity binding assay
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 31.26 kDa, observed mol wt ~40, kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Sustainability features

technique(s)

affinity binding assay: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CD200(4345)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.
OX-2 membrane glycoprotein (UniProt: P41217) is encoded by the CD200 (also known as MOX1, MOX2, My033) gene (Gene ID: 4345) in human. CD200 is a type-1 transmembrane glycoprotein and a member of the immunoglobulin superfamily that functions as a critical immune checkpoint. It is widely expressed on various cell types, including neurons, B cells, activated T cells, and endothelial cells. CD200 is also expressed in the placenta and prevents the mother′s immune system from attacking the fetus. Its primary role is to deliver an immunosuppressive signal by interacting with its receptor, CD200R, which is  predominantly found on myeloid cells like macrophages and microglia. It helps maintain tissue homeostasis by inhibiting the activation of myeloid cells and reducing the production of pro-inflammatory cytokines like TNF-alpha and IL-6. In the central nervous system, neuronal CD200 keeps microglia in a quiescent state. A deficiency in this signaling is linked to chronic neuroinflammation in diseases such as Alzheimer′s and Parkinson′s. CD200 is often overexpressed on the surface of cancer cells (e.g., in chronic lymphocytic leukemia (CLL) and multiple myeloma), acting as a "don′t eat me" signal that allows tumors to evade immune detection. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.
This product aligns with one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with Green Chemistry Principles "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human CD200.

Application

Quality Control Testing
Evaluated by Western Blotting in human placenta tissue lysate. 

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CD200 in human placenta tissue lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CD200 in SK-MEL-28 cell lysate.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CD200 in human placenta tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound with CD200 peptide with at least one thousand-fold (1,000X) higher affinity than with non-specific control peptide.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1F7 is a ZooMAb® rabbit recombinant monoclonal antibody that detects CD200. It targets an epitope within 17 amino acids from the N-terminal half.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library