Skip to Content
MilliporeSigma

ZRB3266

Anti-TOP2B Antibody, clone 5E7-H1 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells

Sustainability features

enhanced validation

Enhanced Validation

Synonym(s):

antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
5E7-H1, recombinant monoclonal
Application:
ICC, IHC, WB, affinity binding assay
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5E7-H1, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 183.27 kDa, observed mol wt ~183 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

rat, nonhuman primates, monkey, boar

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Sustainability features

technique(s)

affinity binding assay: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.
DNA topoisomerase 2-beta (UniProt: Q02880; also known as DNA topoisomerase II, beta isozyme, EC:5.6.2.2) is encoded by the TOP2B gene (Gene ID: 7155) in human. The TOP2B gene is located on human chromosome location 3p24.2. Topoisomerase IIβ (TOP2B) is an isoform of type IIA topoisomerases. It is a member of the topoisomerase II family and is a Key decatenating enzyme that alters DNA topology by binding to two double-stranded DNA molecules, generating a double-stranded break in one of the strands, passing the intact strand through the broken strand, and religating the broken strand. It also plays a role in B-cell differentiation. Structurally, TOP2B consists of an N-terminal domain that facilitates DNA binding and a C-terminal domain that harbors ATPase activity, enabling the enzyme to introduce double-strand breaks in DNA to alleviate torsional strain. TOP2B is predominantly expressed in various tissues, including the brain, heart, and skeletal muscle, with notable localization in the nucleus, where it performs its role in DNA manipulation.  Two isoforms of TOP2B have been described that are produced by alternative splicing. TOP2B has been implicated in neurodegenerative disorders, as its dysfunction can disrupt neuronal homeostasis. Its involvement in disease is particularly significant in cancer, where aberrant expression or mutations in TOP2B can lead to genomic instability and contribute to tumorigenesis. Recent studies have highlighted the potential of TOP2B as a therapeutic target, particularly in the context of chemotherapy, where its inhibitors are used to enhance the efficacy of anticancer agents. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.
We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human TOP2B.

Application

Quality Control Testing
Evaluated by Western Blotting in SH-SY5Y cell lysate. 

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TOP2B in SH-SY5Y cell lysate.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TOP2B in human kidney tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound TOP2B peptide with at least one billion-fold (1,000,000,000X) higher affinity than with non-specific control peptide.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected TOP2B in NIH/3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 5E7-H1 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects TOP2B. It targets an epitope within 20 amino acids from the C-terminal region.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library