Skip to Content
MilliporeSigma

29032

Prothioconazole-(triazole-13C2, 15N3)

analytical standard

Synonym(s):

2-[2-(1-Chlorocyclopropyl)-3-(2-chlorophenyl)-2- hydroxypropyl](13C2,15N3)-2,4-dihydro-3H-1,2,4-triazole-3- thione, [Triazole-13C2, 15N3] Prothioconazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13C2C12H15Cl215N3OS
Molecular Weight:
349.22
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

technique(s)

HPLC: suitable

application(s)

environmental
food and beverages

format

neat

storage temp.

−20°C

SMILES string

ClC1=CC=CC=C1CC(C[15N]2[13C]([15N]=[13CH][15NH]2)=S)(O)C3(Cl)CC3

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Environment

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library