Skip to Content
MilliporeSigma

72733

Bis(1-octyloxy-2,2,6,6-tetramethyl-4-piperidyl) sebacate

analytical standard

Synonym(s):

Bis(1-octyloxy-2,2,6,6-tetramethyl-4-piperidyl) sebacate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C44H84N2O6
CAS Number:
Molecular Weight:
737.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.479 (lit.)

transition temp

Tg >150 °C (dec)

density

1.028 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCON1C(C)(C)CC(CC1(C)C)OC(=O)CCCCCCCCC(=O)OC2CC(C)(C)N(OCCCCCCCC)C(C)(C)C2

InChI

1S/C44H84N2O6/c1-11-13-15-17-23-27-31-49-45-41(3,4)33-37(34-42(45,5)6)51-39(47)29-25-21-19-20-22-26-30-40(48)52-38-35-43(7,8)46(44(9,10)36-38)50-32-28-24-18-16-14-12-2/h37-38H,11-36H2,1-10H3

InChI key

OSIVCXJNIBEGCL-UHFFFAOYSA-N



Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library