Skip to Content
MilliporeSigma

73290

3-Hydroxybenzaldehyde

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein/REAXYS Number:
507099
MDL number:
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

191 °C/50 mmHg (lit.)

mp

100-103 °C (lit.)

application(s)

cosmetics
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

Oc1cccc(C=O)c1

InChI

1S/C7H6O2/c8-5-6-2-1-3-7(9)4-6/h1-5,9H

InChI key

IAVREABSGIHHMO-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 3-Hydroxybenzaldehyde


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library