Skip to Content
MilliporeSigma

74292

Ammonia solution 25%

tested for PFAS methods, LiChropur™

Synonym(s):

Ammonium hydroxide solution, Ammonia aqueous, Ammonia water

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein/REAXYS Number:
3587154
MDL number:
UNSPSC Code:
12352106
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

suitable for EPA 1633, suitable for EPA 533, suitable for EPA 537.1

assay

≥25.0% (acidimetric)

expl. lim.

27 %

technique(s)

HPLC: suitable, LC/MS: suitable, ion chromatography: suitable

density

0.9 g/mL at 25 °C (lit.)

cation traces

Al: ≤5.0 ppb, Ca: ≤20.0 ppb, Fe: ≤20.0 ppb, K: ≤5.0 ppb, Mg: ≤ 5.0 ppb, Na: ≤5.0 ppb

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ammonia solution 25%


Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library