Skip to Content
MilliporeSigma

75299

Isopropyl benzoate

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
MDL number:
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0%

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cosmetics
food and beverages

format

neat

SMILES string

CC(C)OC(=O)c1ccccc1

InChI

1S/C10H12O2/c1-8(2)12-10(11)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

FEXQDZTYJVXMOS-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Isopropyl benzoate


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library