Skip to Content
MilliporeSigma

77495

N,N-Bis(2-hydroxyethyl)dodecanamide

analytical standard

Synonym(s):

Lauric acid diethanolamide

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H33NO3
CAS Number:
Molecular Weight:
287.44
MDL number:
NACRES:
NA.21
eCl@ss:
32039090
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cosmetics
food and beverages

format

neat

storage temp.

2-8°C

SMILES string

N(CCO)(CCO)C(=O)CCCCCCCCCCC

InChI

1S/C16H33NO3/c1-2-3-4-5-6-7-8-9-10-11-16(20)17(12-14-18)13-15-19/h18-19H,2-15H2,1H3

InChI key

AOMUHOFOVNGZAN-UHFFFAOYSA-N



Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library