Skip to Content
MilliporeSigma

77527

Ammonium acetate

tested for PFAS methods, LiChropur

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CO2NH4
CAS Number:
Molecular Weight:
77.08
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32150319
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

suitable for EPA (533), suitable for EPA 1633, suitable for EPA 537.1

assay

≥98.0% (acidimetric)

technique(s)

HPLC: suitable, LC/MS: suitable, ion chromatography: suitable

cation traces

Al: ≤10 ppm, Ca: ≤10 ppm, Fe: ≤10 ppm, K: ≤50 ppm, Mg: ≤10 ppm, Na: ≤50 ppm

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Ammonium acetate


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library